Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bestrophin 1 Antibody (E6-6), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB001848 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001848 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001848 Supplier Novus Biologicals Supplier No. NB300164R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bestrophin 1 Monoclonal specifically detects Bestrophin 1 in Human, Porcine, Canine, Primate, Rat (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Bestrophin 1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone E6-6
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ARB, BEST, Best disease, bestrophin 1, bestrophin-1, BMD, TU15B, vitelliform macular dystrophy 2, Vitelliform macular dystrophy protein 2, VMD2RP50
Gene Symbols BEST1
Host Species Mouse
Immunogen Synthetic peptide conjugated to KLH corresponding to the C-terminus of human Bestrophin 1 (KDHMDPYWALENRDEAHS) [Uniprot: O76090]
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Neuroscience, Sensory Systems, Vision
Primary or Secondary Primary
Gene ID (Entrez) 7439
Test Specificity Bestrophin (E6-6)
Target Species Human, Pig, Canine, Primate, Rat (Negative)
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.