Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-1,3-Glucuronyltransferase 1/B3GAT1 Antibody (3B8B10) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP261708 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61708 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61708 Supplier Novus Biologicals Supplier No. NBP261708
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

beta-1,3-Glucuronyltransferase 1/B3GAT1 Monoclonal specifically detects beta-1,3-Glucuronyltransferase 1/B3GAT1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen beta-1,3-Glucuronyltransferase 1/B3GAT1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 3B8B10
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000
Formulation PBS with 0.05% Sodium Azide
Gene Alias Beta-1,3-glucuronyltransferase 1, beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P), EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, GlcAT-P, GLCATPGLCUATP, glcUAT-P, Glucuronosyltransferase P, NK1, NK-1, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase
Gene Symbols B3GAT1
Host Species Mouse
Immunogen Purified recombinant fragment of human B3GAT1 (AA: 193-334) expressed in E. Coli.
Molecular Weight of Antigen 38.2 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 27087
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.