Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-1,3-Glucuronyltransferase 1/B3GAT1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB338509 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338509 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338509 Supplier Novus Biologicals Supplier No. NBP119788MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

beta-1,3-Glucuronyltransferase 1/B3GAT1 Polyclonal antibody specifically detects beta-1,3-Glucuronyltransferase 1/B3GAT1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen beta-1,3-Glucuronyltransferase 1/B3GAT1
Applications Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Beta-1,3-glucuronyltransferase 1, beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P), EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, GlcAT-P, GLCATPGLCUATP, glcUAT-P, Glucuronosyltransferase P, NK1, NK-1, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human B3TGAT1 protein (between residues 1-50) [UniProt Q9P2W7]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 27087
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.