Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD57 Mouse anti-Human, APC, Clone: TB01, Novus Biologicals™
SDP

Catalog No. NB002341 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002341 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002341 Supplier Novus Biologicals Supplier No. NBP262203APC

Mouse Monoclonal Antibody

CD57 Monoclonal antibody specifically detects CD57 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD57
Applications Flow Cytometry
Classification Monoclonal
Clone TB01
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias HNK1, HNK-1, Human Natural Killer-1, L2, LEU7, LEU-7
Gene Symbols B3GAT1
Host Species Mouse
Immunogen A pool of neuroblastoma cell lines
Purification Method Chromatography
Quantity 0.1 mL
Research Discipline Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 27087
Test Specificity This antibody recognizes CD57, a carbohydrate extracellular antigen present mainly on NK cells, NK T cells, and in neural tissue.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.