Learn More
beta-1,3-Glucuronyltransferase 3/B3GAT3 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | beta-1,3-Glucuronyltransferase 3/B3GAT3 |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 532 |
| Dilution | ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Beta-1,3-glucuronyltransferase 3, beta-1,3-glucuronyltransferase 3 (glucuronosyltransferase I), EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 3, GlcAT-I, GLCATI, GlcUAT-I, Glucuronosyltransferase I, Sqv-8-like protein, UDP-GlcUA:Gal beta-1,3-Gal-R glucuronyltransferase |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant beta-1,3-Glucuronyltransferase 3/B3GAT3 (NP_036332.2; Glu72-Val335). |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.