Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB042182 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB042182 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB042182 Supplier Novus Biologicals Supplier No. NBP297144AF647

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2 Polyclonal specifically detects Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias b3Gal-T7, B3GALT7, B3GNT, B3GNT1, B3GN-T1, B3GN-T2, B3GNT-2, Beta-1,3-galactosyltransferase 7, Beta-1,3-GalTase 7, beta-1,3-Gn-T1, Beta-1,3-Gn-T2, Beta-1,3-N-acetylglucosaminyltransferase 1, Beta-1,3-N-acetylglucosaminyltransferase 2, beta-1,3-N-acetylglucosaminyltransferase bGnT-1, beta-1,3-N-acetylglucosaminyltransferase bGnT-2, Beta3GalT7, Beta3Gal-T7, BETA3GNT, Beta3Gn-T1, beta3Gn-T2, Beta-3-Gx-T7, BGnT-1, BGnT-2, BGNT2, EC 2.4.1.-, UDP-Gal:beta-GlcNAc beta-1,3-galactosyltransferase 7, UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase 7, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 1, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10678
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.