Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-1 Adrenergic R/ADRB1 Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB167271 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167271 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB167271 Supplier Novus Biologicals Supplier No. NBP316089
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

beta-1 Adrenergic R/ADRB1 Polyclonal antibody specifically detects beta-1 Adrenergic R/ADRB1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen beta-1 Adrenergic R/ADRB1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias ADRB1RRHR, adrenergic, beta-1-, receptor, B1AR, beta-1 adrenergic receptor, Beta-1 adrenoceptor, Beta-1 adrenoreceptor, BETA1AR
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 350-450 of human beta-1 Adrenergic R/ADRB1 (NP_000675.1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 153
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.