Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-2 Adrenergic R/ADRB2 Antibody (13D6), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB152951 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB152951 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB152951 Supplier Novus Biologicals Supplier No. NBP312008IR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

beta-2 Adrenergic R/ADRB2 Monoclonal antibody specifically detects beta-2 Adrenergic R/ADRB2 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen beta-2 Adrenergic R/ADRB2
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 13D6
Conjugate DyLight 755
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ADRBR, adrenergic, beta-2-, receptor, surface, B2ARADRB2R, BAR, beta-2 adrenergic receptor, Beta-2 adrenoceptor, Beta-2 adrenoreceptor, BETA2AR, catecholamine receptor
Host Species Rabbit
Immunogen This antibody was raised by immunizing mice with beta-2 adrenergic receptor.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, GPCR, Immunology, Neuroscience, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 154
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.