Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-2 Adrenergic R/ADRB2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB12013989 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12013989 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12013989 Supplier Novus Biologicals Supplier No. NB12013989
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

beta-2 Adrenergic R/ADRB2 Polyclonal specifically detects beta-2 Adrenergic R/ADRB2 in Human samples. It is validated for Western Blot.

Specifications

Antigen beta-2 Adrenergic R/ADRB2
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P07550
Gene Alias ADRBR, adrenergic, beta-2-, receptor, surface, B2ARADRB2R, BAR, beta-2 adrenergic receptor, Beta-2 adrenoceptor, Beta-2 adrenoreceptor, BETA2AR, catecholamine receptor
Gene Symbols ADRB2
Host Species Chicken
Immunogen Synthetic peptide: DFRIAFQELLC LRRSSLKAYGN GYSSNGNTGE QSGYHVEQEKE NKLLCEDLPGT EDFVGHQGTVP SDNIDSQGRNCS T, corresponding to amino acids 335-408 of Human Beta 2 Adrenergic Receptor (NP_000015.1).
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154
Test Specificity Cross-reacts with Human. Not yet tested in other species.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.