Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta 2-Microglobulin Antibody (035), DyLight 405
SDP

Catalog No. NB112223 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB112223 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB112223 Supplier Novus Biologicals Supplier No. NBP290925V

Rabbit Monoclonal Antibody

beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA Detection.

Specifications

Antigen beta 2-Microglobulin
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 35
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Rat beta 2-Microglobulin (P07151; Met1-Met119).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 567
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.