Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta 2-Microglobulin Antibody (246-E9.E7 (same as HLA.ABC.m2)), Novus Biologicals™
SDP

Catalog No. NBP24446501 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24446501 0.1 mg
NBP24446500 0.02 mg
NBP24446502 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP24446501 Supplier Novus Biologicals Supplier No. NBP2444650.1MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence

beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Human, Primate (Negative) samples. It is validated for Flow Cytometry, Immunofluorescence.

Specifications

Antigen beta 2-Microglobulin
Applications Flow Cytometry, SDS-Page, Immunofluorescence
Classification Monoclonal
Clone 246-E9.E7 (same as HLA.ABC.m2)
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml
Formulation 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. P61769
Gene Alias beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin
Gene Symbols B2M
Host Species Mouse
Immunogen Human PBL s from a T-cell acute lymphoblastic leukemia (T-ALL) patient.
Molecular Weight of Antigen 12 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 567
Test Specificity Recognizes a protein of 12kDa, identified as beta-microglobulin. beta-microglobulin non-covalently associates with the 44kDa chain to form the HLA Class I antigen complex. Human beta-2 microglobulin associated with HLA Class I antigens is expressed on many types of cells including lymphocytes, thymocytes, monocytes, granulocytes, platelets, endothelial cells, and epithelial cells. It is absent on erythrocytes. This MAb is specific to human beta-2 microglobulin and does not react with non-human primate cells. This antibody reacts with all cell types excluding erythrocytes. The detection of beta-2 microglobulin in body fluids has been used as a tumor marker and for monitoring patients with HIV infection.
Target Species Human, Primate (Negative)
Content And Storage Store at 4C.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.