Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta 2-Microglobulin Antibody (246-E9.E7 (same as HLA.ABC.m2)), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
| Antigen | beta 2-Microglobulin |
|---|---|
| Clone | 246-E9.E7 (same as HLA.ABC.m2) |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml |
| Applications | Flow Cytometry, SDS-Page, Immunofluorescence |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24446500
![]() |
Novus Biologicals
NBP2444650.02MG |
0.02 mg |
Each for $268.00
|
|
|||||
NBP24446501
![]() |
Novus Biologicals
NBP2444650.1MG |
0.1 mg |
Each for $524.00
|
|
|||||
NBP24446502
![]() |
Novus Biologicals
NBP2444650.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Human, Primate (Negative) samples. It is validated for Flow Cytometry, Immunofluorescence.Specifications
| beta 2-Microglobulin | |
| 0.2 mg/mL | |
| Flow Cytometry, SDS-Page, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
| B2M | |
| IgG2a κ | |
| Protein A or G purified | |
| Recognizes a protein of 12kDa, identified as beta-microglobulin. beta-microglobulin non-covalently associates with the 44kDa chain to form the HLA Class I antigen complex. Human beta-2 microglobulin associated with HLA Class I antigens is expressed on many types of cells including lymphocytes, thymocytes, monocytes, granulocytes, platelets, endothelial cells, and epithelial cells. It is absent on erythrocytes. This MAb is specific to human beta-2 microglobulin and does not react with non-human primate cells. This antibody reacts with all cell types excluding erythrocytes. The detection of beta-2 microglobulin in body fluids has been used as a tumor marker and for monitoring patients with HIV infection. |
| 246-E9.E7 (same as HLA.ABC.m2) | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Primate (Negative) | |
| P61769 | |
| 567 | |
| Human PBL s from a T-cell acute lymphoblastic leukemia (T-ALL) patient. | |
| Primary | |
| Store at 4C. | |
| 12 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title