Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta 2-Microglobulin Antibody (883005), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB373336 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373336 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373336 Supplier Novus Biologicals Supplier No. FAB82481L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen beta 2-Microglobulin
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 883005
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human beta 2-Microglobulin, Met1-Met119, Accession # P61769
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 567
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.