Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta 2-Microglobulin Antibody (B2M/961) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$646.00
Specifications
Antigen | beta 2-Microglobulin |
---|---|
Clone | B2M/961 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Flow (Cell Surface) 1:10-1:1000, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP247704A
![]() |
Novus Biologicals
NBP2477040.1MG |
0.1 mg |
Each for $646.00
|
|
|||||
NBP247704B
![]() |
Novus Biologicals
NBP2477040.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Cell Surface), Immunofluorescence, CyTOF-ready.Specifications
beta 2-Microglobulin | |
1.0 mg/mL | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology | |
PBS with No Preservative | |
567 | |
Full length recombinant human B2M protein | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
12 kDa |
B2M/961 | |
Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Flow (Cell Surface) 1:10-1:1000, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Primate | |
beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
B2M | |
IgG2b κ | |
Protein A or G purified | |
Recognizes a protein of 12kDa, identified as beta-2 microglobulin. Major histocompatibility complex (MHC) class 1 molecules bind to antigens for presentation on the surface of cells. The proteasome is responsible for producing these antigens from the components of foreign pathogens. MHC class 1 molecules consist of an alpha heavy chain that contains three subdomains (alpha1, alpha2, alpha3) and a non-covalent associating light chain, known as beta-2-Microglobulin. beta-2-Microglobulin associates with the alpha3 subdomain of the alpha heavy chain and forms an immunoglobulin domain-like structure that mediates proper folding and expression of MHC class 1 molecules. The alpha1 and alpha2 domains of the alpha heavy chain form the peptide antigen-binding cleft. Mutations in the beta-2-Microglobulin gene can enhance the progression of malignant melanoma phenotypes. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title