Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta 2-Microglobulin Antibody (BBM.1), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB014752 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB014752 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB014752 Supplier Novus Biologicals Supplier No. NBP281045AF532
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen beta 2-Microglobulin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone BBM.1
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin
Gene Symbols B2M
Host Species Rabbit
Immunogen Molt 4, a human T cell line.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 567
Test Specificity Binds human beta 2-Microglobulin, a component of MHC class I molecules, which are present on all nucleated cells (excludes red blood cells) and involved in the presentation of peptide antigens to the immune system.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.