Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta 2-Microglobulin Antibody (SPM617), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N247703PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N247703PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N247703PEC7 Supplier Novus Biologicals Supplier No. NBP247703PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Human, Primate samples. It is validated for Flow Cytometry.

Specifications

Antigen beta 2-Microglobulin
Applications Flow Cytometry
Classification Monoclonal
Clone SPM617
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin
Gene Symbols B2M
Host Species Mouse
Immunogen Full length recombinant human beta 2-Microglobulin protein (Uniprot: P61769)
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 567.0
Test Specificity Beta2 microglobulin is a 12KDa protein with a pI of 5.6. Serum beta2 microglobulin levels are a reflection of cell turnover. Levels rise with fever, inflammation, and infection. Increased serum levels are also seen in B-cell malignancies and in renal failure and may indicate a worse prognosis for patients with early-stage Hodgkin′s lymphoma. In urine, increased levels are seen in proximal renal tubular disease as well as renal transplant rejection. Beta2 microglobulin levels can rise either because its rate of synthesis has increased (e.g. in AIDS, malignant monoclonal plasma cell dyscrasia, solid tumours and autoimmune disease) or because of impaired renal filtration (e.g. due to renal insufficiency, graft rejection or nephrotoxicity induced by post-transplantation immunosuppressive therapy)
Target Species Human, Primate
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.