Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta 2-Microglobulin/FCGRT Antibody (101), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB383105 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383105 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383105 Supplier Novus Biologicals Supplier No. NBP312815V
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

beta 2-Microglobulin/FCGRT Monoclonal antibody specifically detects beta 2-Microglobulin/FCGRT in Human samples. It is validated for ELISA

Specifications

Antigen beta 2-Microglobulin/FCGRT
Applications ELISA
Classification Monoclonal
Clone 101
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias alpha-chain, beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin, Fc fragment of IgG, receptor, transporter, alpha, FcRn, FcRn alpha chain, heavy chain of the major histocompatibility complex class I-like Fc receptor, IgG Fc fragment receptor transporter alpha chain, IgG receptor FcRn large subunit p51, IMD43, immunoglobulin receptor, intestinal, heavy chain, major histocompatibility complex class I-like Fc receptor, Neonatal Fc receptor, neonatal Fc-receptor for Ig, transmembrane alpha chain of the neonatal receptor
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human beta 2-Microglobulin/FCGRT (Uniprot#: P55899-1 & P61770-1; Met1-Ser297 & Met1-Met119).
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 567
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.