Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-3 Adrenergic R/ADRB3 Antibody (493324), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB345723 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345723 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345723 Supplier Novus Biologicals Supplier No. FAB9419MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

beta-3 Adrenergic R/ADRB3 Monoclonal specifically detects beta-3 Adrenergic R/ADRB3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.

Specifications

Antigen beta-3 Adrenergic R/ADRB3
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 493324
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias ADRB3R, adrenergic, beta-3-, receptor, B3AR, beta-3 adrenergic receptor, Beta-3 adrenoceptor, Beta-3 adrenoreceptor, BETA3AR, FLJ99960
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived transfected with human beta-3 Adrenergic R/ADRB3, Met1-Ser408, Accession # P13945
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 155
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.