Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-3 Adrenergic R/ADRB3 Antibody, Novus Biologicals™
SDP

Catalog No. NB10093369 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093369 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10093369 Supplier Novus Biologicals Supplier No. NB10093369
Add to Cart
Add to Cart

Goat Polyclonal Antibody

beta-3 Adrenergic R/ADRB3 Polyclonal specifically detects beta-3 Adrenergic R/ADRB3 in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen beta-3 Adrenergic R/ADRB3
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.03 - 0.1 ug/ml, Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_000016.1
Gene Alias ADRB3R, adrenergic, beta-3-, receptor, B3AR, beta-3 adrenergic receptor, Beta-3 adrenoceptor, Beta-3 adrenoreceptor, BETA3AR, FLJ99960
Gene Symbols ADRB3
Host Species Goat
Immunogen Peptide with sequence PHENSSLAPWPD-C corresponding to N-Terminus according to NP_000016.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 155
Test Specificity Cross reacts with Human. Expected from sequence similarity: Cow.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.