Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-3 Adrenergic R/ADRB3 Antibody, Novus Biologicals™
SDP

Catalog No. NB036450 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036450 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036450 Supplier Novus Biologicals Supplier No. NBP288830
Add to Cart
Add to Cart

Goat Polyclonal Antibody

beta-3 Adrenergic R/ADRB3 Polyclonal specifically detects beta-3 Adrenergic R/ADRB3 in Human samples. It is validated for Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen beta-3 Adrenergic R/ADRB3
Applications Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 5 ug/mL, Peptide ELISA Detection limit 1:32000
Gene Alias ADRB3R, adrenergic, beta-3-, receptor, B3AR, beta-3 adrenergic receptor, Beta-3 adrenoceptor, Beta-3 adrenoreceptor, BETA3AR, FLJ99960
Host Species Goat
Immunogen Peptide with sequence C-EARQSPPLNRFD, from the internal region (near C Terminus) of the protein sequence according to NP_038490.2.
Purification Method Immunogen affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.