Learn More
beta-Actin Antibody, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals NB10056874SS
 View more versions of this product
Catalog No. NB10056874T
Only null left
Add to cart
Description
Description
beta-Actin Polyclonal specifically detects beta-Actin in Human, Mouse, Rat, Porcine, Bovine, Chinese Hamster, Invertebrate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications
Specifications
beta-Actin
Polyclonal
Unconjugated
PBS with 0.05% Sodium Azide
ACTB
Amino acids 2-16 (CDDDIAALVIDNGSG) of actin protein were used as the immunogen sequence for this beta-Actin Antibody.
Protein G purified
RUO
Primary
The NCBI accession number for actin is AAA51567. This can not be used to test heart or muscle lysate loading.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
1.0 mg/mL
Western Blot 0.25-1 ug/ml, Simple Western 1:12.5, Immunohistochemistry reported in scientific literature (PMID 27371029), Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 10 ug/mL, KnockDown Validated reported in scientific literature (PMID 31835509)
ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1
Rabbit
42 kDa
0.025 mg
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers
60
Human, Mouse, Rat, Pig, Invertebrate
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only