Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-Actin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $501.50
Specifications
Antigen | beta-Actin |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 0.25-1 ug/ml, Simple Western 1:12.5, Immunohistochemistry reported in scientific literature (PMID 27371029), Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 10 ug/mL, KnockDown Validated reported in scientific literature (PMID 31835509) |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056874T
![]() |
Novus Biologicals
NB10056874SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056874
![]() |
Novus Biologicals
NB10056874 |
0.1 mg |
Each for $501.50
|
|
|||||
Description
beta-Actin Polyclonal specifically detects beta-Actin in Human, Mouse, Rat, Porcine, Bovine, Chinese Hamster, Invertebrate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
beta-Actin | |
Western Blot 0.25-1 ug/ml, Simple Western 1:12.5, Immunohistochemistry reported in scientific literature (PMID 27371029), Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 10 ug/mL, KnockDown Validated reported in scientific literature (PMID 31835509) | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Pig, Invertebrate | |
ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1 | |
ACTB | |
IgG | |
Protein G purified | |
The NCBI accession number for actin is AAA51567. This can not be used to test heart or muscle lysate loading. |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers | |
PBS with 0.05% Sodium Azide | |
60 | |
Amino acids 2-16 (CDDDIAALVIDNGSG) of actin protein were used as the immunogen sequence for this beta-Actin Antibody. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
42 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title