Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Actin Antibody (8H10D10) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013668 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013668 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013668 Supplier Novus Biologicals Supplier No. NBP280595
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

beta-Actin Monoclonal specifically detects beta-Actin in Human, Mouse, Rat, Chinese Hamster, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoblotting, CyTOF-ready.

Specifications

Antigen beta-Actin
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoblot, CyTOF
Classification Monoclonal
Clone 8H10D10
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1:1000-1:5000, Simple Western : 12.5, Flow Cytometry : 1:200-1:400, ELISA : 1:10000, Immunocytochemistry/Immunofluorescence : 1:200-1:1000, Immunoblotting reported in scientific literature (PMID 27903531), CyTOF-ready
Formulation PBS
Gene Alias ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1
Gene Symbols ACTB
Host Species Mouse
Immunogen This beta-Actin Antibody (8H10D10) was made to a synthetic peptide corresponding to amino-terminal residues of human Beta Actin, conjugated to KLH. [UniProt# P60709]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 60
Target Species Human, Mouse, Rat, Chinese Hamster, Primate
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.