Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Actin Antibody (AC-15), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB170402 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170402 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB170402 Supplier Novus Biologicals Supplier No. NB11067828AF750
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

beta-Actin Monoclonal antibody specifically detects beta-Actin in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Feline, Fish, Guinea Pig, Hamster, Leech, Mammalia, Primate, Rabbit, Sheep, Squirrel, Turkey, Zebrafish, Drosophila (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen), KnockDown, KnockDown

Specifications

Antigen beta-Actin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen), KnockDown, KnockDown
Classification Monoclonal
Clone AC-15
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1
Host Species Mouse
Immunogen This beta-Actin Antibody (AC-15) was made to a slightly modified Beta-cytoplasmic actin N-terminal peptide, Ac-Asp-Asp-Asp-Ile-Ala-Ala-Leu-Val-Ile-Asp-Asn-Gly-Ser-Gly-Lys, conjugated to KLH.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 60
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Feline, Fish, Guinea Pig, Hamster, Leech, Mammalia, Primate, Rabbit, Sheep, Squirrel, Turkey, Zebrafish, Drosophila (Negative)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.