Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Actin Antibody (AC-15), Novus Biologicals™
SDP

Catalog No. NB600501 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600501 0.1 mL
NB600501SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB600501 Supplier Novus Biologicals Supplier No. NB600501
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 442 publications

beta-Actin Monoclonal specifically detects beta-Actin in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Feline, Fish, Guinea Pig, Hamster, Leech, Mammal, Primate, Rabbit, Sheep, Squirrel, Turkey, Zebrafish, Drosophila (Negative) samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Chromatin Immunoprecipitation (ChIP), Knockout Validated, Knockdown Validated.

Specifications

Antigen beta-Actin
Applications ChIP Assay, ELISA, Flow Cytometry, Immunoassay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown
Classification Monoclonal
Clone AC-15
Conjugate Unconjugated
Dilution Western Blot 1:5000 - 1:10000, Simple Western 1:25, Flow Cytometry, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:2000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:500, Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated
Gene Accession No. P60709
Gene Alias ACTB, actin, beta, actin, cytoplasmic 1, anti-beta actin, B-Actin, Beta Actin, beta actin (ac-15), beta cytoskeletal actin, Beta-actin, beta-actin ac 15, beta-actin ac15, beta-actin ac-15, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1
Gene Symbols ACTB
Host Species Mouse
Immunogen This beta-Actin Antibody (AC-15) was made to a slightly modified Beta-cytoplasmic actin N-terminal peptide, Ac-Asp-Asp-Asp-Ile-Ala-Ala-Leu-Val-Ile-Asp-Asn-Gly-Ser-Gly-Lys, conjugated to KLH.
Molecular Weight of Antigen 42 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 60
Test Specificity In staining of chicken gizzard ultrathin tissue cryosections, the antibody labels the dense bodies and longitudinal channels linking consecutive dense bodies that are also occupied by desmin and the membrane-associated dense plaque. It does not stain adult cardiac and skeletal muscles except for traces due to contaminations of the sample with non-muscle cells, or if embryonic tissue is being used. The epitope recognized by the antibody is resistant to formalin-fixed and paraffin-embedding.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Feline, Fish, Guinea Pig, Hamster, Leech, Mammalia, Primate, Rabbit, Sheep, Squirrel, Turkey, Zebrafish, Drosophila (Negative)
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.