Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-Actin Antibody (AC-15), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 442 publications
$658.00
Specifications
Antigen | beta-Actin |
---|---|
Clone | AC-15 |
Dilution | Western Blot 1:5000 - 1:10000, Simple Western 1:25, Flow Cytometry, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:2000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:500, Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
beta-Actin Monoclonal specifically detects beta-Actin in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Feline, Fish, Guinea Pig, Hamster, Leech, Mammal, Primate, Rabbit, Sheep, Squirrel, Turkey, Zebrafish, Drosophila (Negative) samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Chromatin Immunoprecipitation (ChIP), Knockout Validated, Knockdown Validated.Specifications
beta-Actin | |
Western Blot 1:5000 - 1:10000, Simple Western 1:25, Flow Cytometry, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:2000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:500, Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated | |
Unconjugated | |
Mouse | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers | |
ACTB, actin, beta, actin, cytoplasmic 1, anti-beta actin, B-Actin, Beta Actin, beta actin (ac-15), beta cytoskeletal actin, Beta-actin, beta-actin ac 15, beta-actin ac15, beta-actin ac-15, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1 | |
ACTB | |
IgG1 | |
Unpurified | |
In staining of chicken gizzard ultrathin tissue cryosections, the antibody labels the dense bodies and longitudinal channels linking consecutive dense bodies that are also occupied by desmin and the membrane-associated dense plaque. It does not stain adult cardiac and skeletal muscles except for traces due to contaminations of the sample with non-muscle cells, or if embryonic tissue is being used. The epitope recognized by the antibody is resistant to formalin-fixed and paraffin-embedding. |
AC-15 | |
Monoclonal | |
Ascites | |
RUO | |
P60709 | |
60 | |
This beta-Actin Antibody (AC-15) was made to a slightly modified Beta-cytoplasmic actin N-terminal peptide, Ac-Asp-Asp-Asp-Ile-Ala-Ala-Leu-Val-Ile-Asp-Asn-Gly-Ser-Gly-Lys, conjugated to KLH. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
42 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title