Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Actin Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB120787 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120787 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120787 Supplier Novus Biologicals Supplier No. NB600505JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

beta-Actin Polyclonal antibody specifically detects beta-Actin in Human, Mouse, Rat, Amphibian samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence

Specifications

Antigen beta-Actin
Applications ELISA, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1
Host Species Rabbit
Immunogen beta-Actin Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to C-Terminal region near amino acids 350-375 of Human beta Actin. (Uniprot: P60709)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 60
Target Species Human, Mouse, Rat, Amphibian
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.