Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Actin Antibody, Novus Biologicals™
SDP

Catalog No. NB600505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600505 0.1 mg
NB600505SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB600505 Supplier Novus Biologicals Supplier No. NB600505
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 13 publications

beta-Actin Polyclonal specifically detects beta-Actin in Human, Mouse, Rat, Amphibian samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Electron Microscopy.

Specifications

Antigen beta-Actin
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:4000, ELISA 1:10000-1:40000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:500-1:2000, Immunohistochemistry-Paraffin 1:50 - 1:200, Electron Microscopy
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. P60709
Gene Alias ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1
Gene Symbols ACTB
Host Species Rabbit
Immunogen beta-Actin Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to C-Terminal region near amino acids 350-375 of Human beta Actin. (Uniprot: P60709)
Molecular Weight of Antigen 42 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 60
Test Specificity Expected to cross-react with a wide range of species including most vertebrates and some yeast due to sequence identity.
Target Species Human, Mouse, Rat, Amphibian, Bovine, Guinea Pig, Goat, Hamster, Sheep
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.