Learn More
Description
Specifications
Specifications
| Antigen | beta-Actin |
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | PE |
| Dilution | Flow Cytometry |
| Formulation | PBS |
| Gene Alias | ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1 |
| Gene Symbols | ACTB |
| Host Species | Rabbit |
| Immunogen | This beta-Actin Antibody was made from a synthetic peptide made to an N-terminal region of human Beta Actin. [UniProt P60709] |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
