Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Actin Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991842
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1842 200 μg
50-199-1843 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1842 Supplier Rockland Immunochemicals Supplier No. 600401886
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-beta-Actin Loading Control Antibody reacts with Human, Rat, Monkey and Mouse. beta-Actin Loading Control Antibody is expected to cross-react with a wide range of species due to sequence homology. Anti-Beta Actin is affinity-purified antibody is directed against human beta Actin protein. A BLAST analysis was used to suggest that this antibody would react with beta Actin from a wide range of organisms, including most vertebrates and some yeast. Broad reactivity makes this antibody an excellent loading control.

Beta actin is one of six actin isoforms which have been identified. Beta actin is a non-muscle cytoskeletal protein in all human cell types and is involved in cell motility, structure, and integrity. There are six different actin isoforms in human. The beta isoform of actin, along with gamma actin, coexist in most cell types as components of the cyto-skeleton. Beta actins are cytoplasmic proteins ubuquitously expressed in all eukaryotic cells. Polymerization of globular actin (G-actin) leads to a structural filament (F-actin) in the form of two stranded helix. Actins are highly conserved proteins that are involved in cell motility, structure and integrity. Because beta actin is ubiquitously expressed in all eukaryotic cells, it is frequently used as a loading control for assays involving protein detection, such as Western blotting. Antibodies to beta-actin provide a specific and useful tool in studying the intracellular distribution of beta-actin and the static and dynamic aspects of the cytoskeleton.

Specifications

Antigen beta Actin
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene ACTB
Gene Accession No. P48975, P60709, P60710, P60711, P60712, P60713, Q6QAQ1, Q71FK5
Gene Alias ACT-5; ACTA1; ACTB; actb.L; actb1; actba; actin; actin beta; actin, alpha 1, skeletal muscle; actin, beta; actin, beta 1; actin, beta L homeolog; actin, beta, cytoplasmic; Actin, cytoplasmic 1; Actin, cytoplasmic 1, N-terminally processed; Actx; Atcb; bact; bactin1; bactin1 protein; bactzf; B-ACTZF; beta actin; beta cytoskeletal actin; Beta-actin; Beta-actin-1; BOS_22932; BRWS1; Cap Z; capZ beta; cytoplasmic 1; Cytoplasmic beta-actin; cytoskeletal protein; cytoskeleton actin; E430023M04Rik; gamma actin; gamma non-muscle actin; PS1TP5-binding protein 1; PS1TP5BP1; RCJMB04_4h19; wu:fd18f05; XELAEV_18045052mg
Gene Symbols ACTB
Host Species Rabbit
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100135470, 100689477, 101844587, 102179831, 11461, 280979, 414396, 443052, 60, 81822
Target Species Bovine, Goat, Guinea Pig, Hamster, Human, Monkey, Mouse, Ovine, Pig, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.