Learn More
beta Actin Rabbit anti-Human, Mouse, Non-human primate, Rat, Zebrafish, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
This antibody was generated against N-terminal region of human beta actin protein and can cross-react with other actins.
Specifications
Specifications
| Antigen | beta Actin |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.43 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ACTA1 |
| Gene Accession No. | P60709, P60710, P60711, P62736, P62737, P62738, P63259, P63260, P63261, P63267, P63268, P63269, P68032, P68033, P68035, P68133, P68134, P68136 |
| Gene Alias | ACTB, Actin, actin, beta, Actin, cytoplasmic 1, B actin, Beta actin, F actin, PS1TP5BP1, beta actin |
| Gene Symbols | Acta1, acta1b, acta2, ACTB, actb1, Actc1, actc1b, Actg1, Actg2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.