Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Alanine methyl ester hydrochloride, 98%

Catalog No. AAH2748306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2748306 5 g
AAH2748314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2748306 Supplier Thermo Scientific Chemicals Supplier No. H2748306
Only null left
Add to Cart

CAS: 3196-73-4 | C4H10ClNO2 | 139.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3196-73-4
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00039060
InChI Key XPGRZDJXVKFLHQ-UHFFFAOYSA-N
Synonym methyl 3-aminopropionate hydrochloride, methyl 3-aminopropanoate hydrochloride, beta-alanine methyl ester hydrochloride, beta-alanine methyl ester hcl, h-beta-ala-ome.hcl, h-, a-ala-ome.hcl, h-beta-ala-ome hydrochloride, b-alanine methyl ester hydrochloride, 3-aminopropanoic acid methyl ester hydrochloride, 3-aminopropionic acid methyl ester hydrochloride
PubChem CID 2734767
IUPAC Name methyl 3-aminopropanoate;hydrochloride
SMILES Cl.COC(=O)CCN

Specifications

Melting Point 102°C to 104°C
Linear Formula H2NCH2CH2CO2CH3·HCl
Beilstein 3556748
Quantity 5 g
Formula Weight 139.58
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material beta-Alanine methyl ester hydrochloride
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.