Learn More
Description
Specifically detects beta Amyloid 9 in Human, Mouse, Primate, Rat samples, and it is validated for ELISA, Western Blotting
Specifications
Specifications
| Antigen | beta Amyloid 9 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Concentration | 1.32 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1ug/ml, ELISA 1:10000 |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 50% (v/v) Glycerol with 0.01% Sodium Azide |
| Gene Accession No. | P05067, P05067, P05067, P05067 |
| Gene Alias | ABPP, Alzheimer disease amyloid protein, APP1, Cerebral vascular amyloid peptide, Protease nexin-II |
| Host Species | Sheep |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
