Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Amyloid Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB223112 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB223112 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB223112 Supplier Novus Biologicals Supplier No. NBP225093AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

beta Amyloid Polyclonal antibody specifically detects beta Amyloid in Human, Mouse, Rat, Bacteria samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen beta Amyloid
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias Abeta, Amyloid beta, beta-amyloid peptide
Host Species Rabbit
Immunogen This beta Amyloid antibody was raised against a 10 amino acid peptide on the amino terminus of the 4KDa beta Amyloid peptide generated by beta- and gamma-secretases. The immunogen is located within amino acids 650 - 700 of human amyloid A4 protein precursor (APP). Amino Acid Squence: DAEFRHDSGYE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cardiovascular Biology, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 351
Target Species Human, Mouse, Rat, Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.