Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Amyloid Antibody (MOAB-2), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP213075C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP213075C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP213075C Supplier Novus Biologicals Supplier No. NBP213075C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

beta Amyloid Monoclonal specifically detects beta Amyloid in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Immunohistochemistry Free-Floating.

Specifications

Antigen beta Amyloid
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunohistochemistry (Free Floating)
Classification Monoclonal
Clone MOAB-2
Conjugate DyLight 650
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Immunohistochemistry Free-Floating
Gene Accession No. P05067
Gene Alias Abeta, Amyloid beta, beta-amyloid peptide
Gene Symbols APP
Host Species Mouse
Immunogen This beta Amyloid antibody was developed against recombinant human beta Amyloid 42.
Molecular Weight of Antigen 5 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cardiovascular Biology, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 351
Test Specificity Beta Amyloid antibody (MOAB-2) recognizes unaggregated, oligomeric and fibrillar forms of beta Amyloid 42 and unaggregated beta Amyloid 40. Does not detect APP.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.