Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Arrestin 2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP224569SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224569SS 0.025 mg
NBP224569 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224569SS Supplier Novus Biologicals Supplier No. NBP224569SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

beta-Arrestin 2 Polyclonal specifically detects beta-Arrestin 2 in Human, Mouse, Rat, Bovine, Canine, Chicken, Equine, Primate, Xenopus, Zebrafish samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen beta-Arrestin 2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.25 - 0.5 ug/mL, Simple Western 1:100, Immunohistochemistry-Paraffin 5 ug/mL
Formulation PBS with 0.05% Sodium Azide
Gene Alias ARB2, ARR2, arrestin 3, Arrestin beta-2, arrestin, beta 2, BARR2, beta-arrestin-2, DKFZp686L0365
Gene Symbols ARRB2
Host Species Rabbit
Immunogen A peptide sequence corresponding to an area between amino acids 1 and 50 of human ARRB2 was used as the immunogen for the antibody, Gen Bank no. gb:ABG47460.1.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 409
Test Specificity The eIF3A amino acid sequence used as immunogen for the antibody is 100% homologous in human, rat, mouse, Xenopus, chicken, chimpanzee, zebrafish, cow, platypus, horse, opossum and boar, and 94% homologous in Tetradon.
Target Species Human, Mouse, Rat, Bovine, Canine, Chicken, Equine, Primate, Xenopus species, Zebrafish
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.