Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Casein Antibody (F20.14) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB1002720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002720 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002720 Supplier Novus Biologicals Supplier No. NB1002720
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

beta Casein Monoclonal specifically detects beta Casein in Human samples. It is validated for Immunohistochemistry, Western Blot.

Specifications

Antigen beta Casein
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone F20.14
Concentration 0.88 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Gene Alias beta-casein, CASB, casein beta
Gene Symbols CSN2
Host Species Mouse
Immunogen Purified casein and a-lactalbumin.
Molecular Weight of Antigen 28 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1447
Test Specificity Does not cross react with alpha-lactalbumin.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.