Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Beta-casein Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $525.50
Specifications
Antigen | Beta-casein |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5118998
|
Invitrogen™
PA5118998 |
200 μL |
Each for $525.50
|
|
|||||
PIPA5122201
|
Invitrogen™
PA5122201 |
20 μL |
Each for $209.50
|
|
|||||
Description
Beta-casein Polyclonal Antibody for Western Blot, IHC (P)
This gene is a member of the beta casein family. There are two types of casein protein, beta (encoded by this gene) and kappa, both of which are secreted in human milk. Beta casein is the principal protein in human milk and the primary source of essential amino acids for a suckling infant. Beta and kappa casein proteins acting together form spherical micelles which bind within them important dietary minerals, such as calcium and phosphorous. Alternative splicing results in multiple transcript variants.Specifications
Beta-casein | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Mouse, Rat | |
CSN2 | |
beta casein; beta-caesin; beta-casein; beta-casein A3; beta-casein precursor; CASB; casein beta; CSN2; kappa-casein; Unknown (protein for MGC:133765); unnamed protein product | |
CSN2 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P02665, P05814, P10598 | |
12991, 1447, 29173 | |
Recombinant protein Casein Beta. The antigen corresponds to amino acid range 41-198 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title