Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Catenin Antibody (12F7) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP154467SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP154467SS 0.025 mL
NBP154467 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP154467SS Supplier Novus Biologicals Supplier No. NBP154467SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 13 publications

beta-Catenin Monoclonal specifically detects beta-Catenin in Human, Mouse, Rat, Duck, Chicken, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular).

Specifications

Antigen beta-Catenin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 12F7
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western 1:100, Flow Cytometry, Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200, Flow (Intracellular)
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P35222, Q02248, Q9WU82, O42486
Gene Alias beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923
Gene Symbols CTNNB1
Host Species Mouse
Immunogen Recombinant chicken beta Catenin fused to maltose binding protein. [UniProt# O42486]
Molecular Weight of Antigen 92 kDa
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Extracellular Matrix, Neuroscience, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1499
Target Species Human, Mouse, Rat, Duck, Chicken, Primate
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.