Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Catenin Antibody (12F7) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 13 publications

$227.50 - $568.50

Specifications

Antigen beta-Catenin
Clone 12F7
Concentration 1.0 mg/mL
Dilution Western Blot 1:1000, Simple Western 1:100, Flow Cytometry, Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200, Flow (Intracellular)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP154467SS
SDP
View Documents
Novus Biologicals
NBP154467SS
0.025 mL
Each for $227.50
Only null left
Add to Cart
 
NBP154467
SDP
View Documents
Novus Biologicals
NBP154467
0.1 mL
Each for $568.50
Only null left
Add to Cart
 
Description

Description

beta-Catenin Monoclonal specifically detects beta-Catenin in Human, Mouse, Rat, Duck, Chicken, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular).
Specifications

Specifications

beta-Catenin
1.0 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer, Cellular Markers, Extracellular Matrix, Neuroscience, Stem Cell Signaling Pathway, Wnt Signaling Pathway
PBS with 0.02% Sodium Azide
beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923
CTNNB1
IgG1
Protein G purified
92 kDa
12F7
Western Blot 1:1000, Simple Western 1:100, Flow Cytometry, Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200, Flow (Intracellular)
Monoclonal
Purified
RUO
Human, Mouse, Rat, Duck, Chicken, Primate
P35222, Q02248, Q9WU82, O42486
1499
Recombinant chicken beta Catenin fused to maltose binding protein. [UniProt# O42486]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.