Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Catenin Antibody (CTNNB1/1508), Novus Biologicals™
SDP

Catalog No. NB253365200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB253365200 200 μg
NB25336520 20 μg
NB253365100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB253365200 Supplier Novus Biologicals Supplier No. NBP253365200UG

Mouse Monoclonal Antibody

beta-Catenin Monoclonal antibody specifically detects beta-Catenin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen beta-Catenin
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CTNNB1/1508
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml
Formulation 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Gene Alias beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923
Gene Symbols CTNNB1
Host Species Mouse
Immunogen Recombinant human full-length beta-Catenin protein fragment
Molecular Weight of Antigen 92 kDa
Purification Method Protein A or G purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Extracellular Matrix, Mucosal Immunology, Neuroscience, Phospho Specific, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1499
Target Species Human, Mouse, Rat
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.