Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta COP Antibody, Novus Biologicals™
SDP

Catalog No. NBP222544 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222544 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222544 Supplier Novus Biologicals Supplier No. NBP222544
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

beta COP Polyclonal specifically detects beta COP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen beta COP
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 4.4 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:2000
Formulation PBS containing 1 mg/ml BSA with 0.05% Sodium Azide
Gene Accession No. 70349, 114023, P53618, 70349, 114023
Gene Alias beta coat protein, Beta-coat protein, beta-cop, coatomer protein complex, subunit beta, coatomer protein complex, subunit beta 1, coatomer subunit beta, COPBFLJ57957, DKFZp761K102, FLJ10341, FLJ46444
Gene Symbols COPB1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues E(496) A G E L K P E E E I T V G P V Q K(513) of rat beta-COP.
Purification Method Ammonium sulfate precipitation
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1315
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.