Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Cyclocitral, 90+%

Catalog No. p-7028405
Click to view available options
Quantity:
10 g
50 g

CAS: 432-25-7 | C10H16O | 152.24 g/mol

beta-Cyclocitral is used as flavoring agent, nutrient, stabilizers, surfactants and emulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
beta-Cyclocitral is used as flavoring agent, nutrient, stabilizers, surfactants and emulsifiers.

Solubility
Soluble in chloroform, dichloromethane, ethyl acetate and methanol. Insoluble in water.

Notes
Air sensitive. Store away from air, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 432-25-7
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00079078
InChI Key MOQGCGNUWBPGTQ-UHFFFAOYSA-N
Synonym beta-cyclocitral, b-cyclocitral, 1-cyclohexene-1-carboxaldehyde, 2,6,6-trimethyl, .beta.-cyclocitral, 2,6,6-trimethyl-1-cyclohexene-1-carboxaldehyde, unii-77y0u2x29g, cyclocitral, alpha beta-cyclocitral, 2,6,6-trimethylcyclohex-1-ene-1-carbaldehyde, 1-formyl-2,6,6-trimethyl-1-cyclohexene
PubChem CID 9895
ChEBI CHEBI:53177
IUPAC Name 2,6,6-trimethylcyclohexene-1-carbaldehyde
SMILES CC1=C(C=O)C(C)(C)CCC1

Specifications

Density 0.944
Boiling Point 62°C to 63°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.498
Quantity 10 g
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform,dichloromethane,ethyl acetate and methanol. Insoluble in water.
Formula Weight 152.23
Percent Purity ≥90%
Chemical Name or Material beta-Cyclocitral
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-081-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.