Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Defensin 3 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB181826 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB181826 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB181826 Supplier Novus Biologicals Supplier No. NBP306517AF488
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

beta-Defensin 3 Polyclonal antibody specifically detects beta-Defensin 3 in Human samples. It is validated for ELISA, Immunofluorescence

Specifications

Antigen beta-Defensin 3
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias BD3, BD-3, beta-defensin 103, Beta-defensin 3, DEFB103, DEFB-3, DEFB3DEFB103A, Defensin, beta 103, defensin, beta 103B, Defensin-like protein, hBD-3, HBD3HBP3
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human beta-Defensin 3 (P81534; Gly23-Lys6).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 55894
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.