Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Defensin 3 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB003961 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003961 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003961 Supplier Novus Biologicals Supplier No. NB200117AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

beta-Defensin 3 Polyclonal antibody specifically detects beta-Defensin 3 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).

Specifications

Antigen beta-Defensin 3
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BD3, BD-3, beta-defensin 103, Beta-defensin 3, DEFB103, DEFB-3, DEFB3DEFB103A, Defensin, beta 103, defensin, beta 103B, Defensin-like protein, hBD-3, HBD3HBP3
Gene Symbols DEFB103B
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of Defensin beta 3 (between amino acids 10-60) [UniProt P81534]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Immunology
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.