Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Galactosidase Antibody (13R4), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB014485 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB014485 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB014485 Supplier Novus Biologicals Supplier No. NBP281034AF488
Add to Cart
Add to Cart

Rat Monoclonal Antibody

beta Galactosidase Monoclonal specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen beta Galactosidase
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 13R4
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias EC 3.2.1.23; b-gal
Host Species Rat
Immunogen beta Galactosidase.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity 13R4 recognizes wild-type E. coli beta-galactosidase.
Target Species E. coli
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.