Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Galactosidase Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB001822 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1000 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001822 1000 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001822 Supplier Novus Biologicals Supplier No. NB12066411000UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

beta Galactosidase Polyclonal specifically detects beta Galactosidase in Mouse, E. coli samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.

Specifications

Antigen beta Galactosidase
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate FITC
Dilution Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:500-1:2500, Fluorophore-linked immunosorbent assay 1:100-1:2000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free
Gene Alias EC 3.2.1.23; b-gal
Host Species Rabbit
Immunogen Full length native Beta Galactosidase isolated from E.coli.
Quantity 1000 μg
Research Discipline Epitope Tags, Fusion Proteins
Test Specificity Beta-Galactosidase
Target Species E. coli
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.