Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Galactosidase Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB358431 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358431 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358431 Supplier Novus Biologicals Supplier No. NBP176322MFV610

Chicken Polyclonal Antibody

beta Galactosidase Polyclonal antibody specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot, ELISA

Specifications

Antigen beta Galactosidase
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 3.2.1.23; b-gal
Host Species Chicken
Immunogen Antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of E. coli beta-gal. The immunogen is located within amino acids 20 - 70 of beta-Galactosidase. Amino Acid Squence: WRNSEEARTDRPSQQLR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species E. coli
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.