Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Galactosidase Antibody, Novus Biologicals™
SDP

Catalog No. NBP242819 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242819 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242819 Supplier Novus Biologicals Supplier No. NBP242819
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects beta Galactosidase in Other samples, and it is validated for Western Blotting

beta Galactosidase Polyclonal specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot.

Specifications

Antigen beta Galactosidase
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Alias EC 3.2.1.23; b-gal
Host Species Rabbit
Immunogen The immunogen used to generate this antibody corresponds to Beta-gal
Molecular Weight of Antigen 116 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species E. coli
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.