Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta I Tubulin Antibody (16E2), Novus Biologicals™
SDP

Catalog No. NB298442 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298442 50 μL
NB298443 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298442 Supplier Novus Biologicals Supplier No. NBP32626750UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

beta I Tubulin Monoclonal antibody specifically detects beta I Tubulin in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry

Specifications

Antigen beta I Tubulin
Applications Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 16E2
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200
Formulation 50% Glycerol, 0.01M PBS, PH 7.4
Gene Alias beta tubulin 1, class VI, dJ543J19.4, tubulin beta-1 chain, tubulin, beta 1
Host Species Mouse
Immunogen Recombinant Human beta I Tubulin protein [UniProt Q9H4B7]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 81027
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.